Recombinant Rat GM-CSF control protein
- Control Protein
- PCA Assays
- ImmunoBlot
Granulocyte Macrophage Colony Stimulating Factor (GM-CSF) is hematopoietic factor produced by entothelial cells, monocytes, fibroblasts and T cells in response to a number of inflammatory mediators. GM-CSF is able to stimulate the production of neutrophilic granulocytes, macrophages, and mixed granulocyte-macrophage colonies from bone marrow cells. GM-CSF can also stimulate some functional activities in mature granulocytes and macrophages. Human and mouse GM-CSF show no cross-reactivity. Recombinant rat GM-CSF is a non-glycosylated protein, containing 128 amino acids, with a molecular weight of 14.6 kDa.
Granulocyte Macrophage Colony Stimulating Factor Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Granulocyte Macrophage Colony Stimulating Factor in immunological assays.