Would you like to log in again?
rMouse IL-33 Protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
010F01V68-0010 010F01V68-1000 010F01V68-0100 010F01V68-0002
75934-744 75934-742 75934-746 75934-748
rMouse IL-33 Protein, Rockland Immunochemicals
Recombinant Mouse IL-33 control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Interleukin 33 (IL-33) is a member of the IL-1 cytokine family and is constitutively expressed in smooth muscle cells and the airway epithelium. IL-33 signals through receptors, IL-1RL1 and IL-1RAP, to induce type 2 cytokine production in polarized Th2 cells. Recombinant mouse IL-33 is a non-glycosylated protein, containing 159 amino acids, with a molecular weight of 17.7 kDa.
Interleukin-33 Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Interleukin-33 in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty