Would you like to log in again?
rMouse IL-15 Protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
010F01V53-0010 010F01V53-0100 010F01V53-1000 010F01V53-0002
75934-680EA 293.94 USD InStock
75934-680 75934-682 75934-678 75934-684
rMouse IL-15 Protein, Rockland Immunochemicals
Proteins and Peptides
Recombinant Mouse IL-15 control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Interleukin 15 (IL-15) is a widely expressed pro-inflammatory cytokine related to IL-2. IL-15 promotes activation of T cells, neutrophils and macrophages and is critical to dendritic cell function in several model systems. IL-15 has been shown to play a role in several inflammatory disorders, including rheumatoid arthritis, psoriasis and pulmonary inflammatory diseases. Emerging data suggest that IL-15 may serve as a good therapeutic target, as there appears to be a beneficial effect of IL-15 neutralization in models of psoriasis and diabetes. Human IL-15 shows activity on mouse cells. Recombinant mouse IL-15 is a non-glycosylated protein, containing 115 amino acids, with a molecular weight of 13.3 kDa.
Interleukin-15 Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Interleukin-15 in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty