Would you like to log in again?
rMouse IL-4 Protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
010F01B93-1000 010F01B93-0005 010F01B93-0100 010F01B93-0020
75934-760EA 5549.89 USD InStock
75934-760 75929-164 75929-156 75929-160
rMouse IL-4 Protein, Rockland Immunochemicals
Proteins and Peptides
Recombinant Mouse IL-4 control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Interleukin-4 (IL-4) is an immuno-modulatory cytokine, produced primarily by a subgroup of helper T cells, known as Th2. IL-4 is known to induce naïve helper T cells to differentiate to Th2 cells, which subsequently produce more IL-4. Th2 cells regulate a wide variety of other immune cells including, B cells, where it has the ability to promote IgG to IgE isotype switching. Recombinant mouse IL-4 is a non-glycosylated protein, containing 121 amino acids, with a molecular weight of 13.5 kDa.
Interleukin-4 Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Interleukin-4 in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty