Mouse IgG2b Isotype Control (RPE)
Supplier: Rockland Immunochemicals
010-0842
75934-902EA 329.1 USD
75934-902
Mouse IgG2b Isotype Control (RPE)
Antibodies
Isotype controls are important for Flow Cytometry and have no specificity for target cells within a particular experiment. Their purpose is to confirm the specificity of primary antibody binding that it is not a result of non-specific Fc receptor binding to cells or other cellular protein interactions. Isotype controls need to be matched to the specific primary Abs (species and isotype, including heavy and light chains) being used.
Mouse IgG2b isotype control can be utilized as a control or standard reagent in Flow cytometry, Western Blotting, and ELISA experiments where determination of sample isotype is important.
Applications: control or standard reagent, Flow cytometry, Western Blotting, and ELISA