Would you like to log in again?
rMouse PDGF-AA Protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
010001W08-0010 010001W08-0100 010001W08-0002 010001W08-1000
75931-062 75931-064 75931-066 75931-060
rMouse PDGF-AA Protein, Rockland Immunochemicals
Recombinant Mouse PDGF-AA control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Platelet-Derived Growth Factor (PDGF) is a mitogenic peptide growth hormone carried in the alpha-granules of platelets and is released when platelets adhere to traumatized tissues. Connective tissue cells near the traumatized region respond by initiating the process of replication. The synthesis of PDGF can be induced by IL-1, IL-6, TNF-α, TGF-β and EGF. Recombinant mouse PDGF-AA is a non-glycosylated, disulfide-linked homodimer, containing two 126 amino acid chains, with a total molecular weight of 28.9 kDa.
Platelet Derived Growth Factor-AA Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Platelet Derived Growth Factor-AA in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty