Would you like to log in again?
rMouse Neurotrophin-3 Protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
010001W06-0100 010001W06-0002 010001W06-1000 010001W06-0010
75933-996EA 834.85 USD InStock
75933-996 75934-000 75933-988 75933-992
rMouse Neurotrophin-3 Protein, Rockland Immunochemicals
Proteins and Peptides
Recombinant Mouse Neurotrophin-3 control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Neurotrophin-3 (NT-3) is an important member of the nerve growth factor (NGF) family of proteins behind BDNF and NGF. It is thought to promote the survival or differentiation of existing and new neurons in the central nervous system and synapses. These functions are thought to be communicated by TrkC, a receptor tryrosine kinase thought to induce NT-3-specific signaling. In addition, NT-3 is thought to also bind TrkB and low affinity nerve growth factor receptor (LNGFR). Recombinant mouse NT-3 is a non-glycosylated, non-covalently linked homodimer, containing two 119 amino acid chains, with a total molecular weight of 27.2 kDa.
Neurotrophin-3 Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Neurotrophin-3 in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty