Mouse IL-21
Supplier: Rockland Immunochemicals
010-001-V61-1000 010-001-V61-0010 010-001-V61-0100 010-001-V61-0002
75929-142EA 960.74 USD
75929-142 75928-794 75929-134 75929-138
Mouse IL-21
Proteins and Peptides
Recombinant Mouse IL-21 control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Interleukin-21 (IL-21) is a member of the common gamma-chain family of cytokines (includes IL-2, IL-4, IL-7, IL-9, IL-13, and IL-15) and is expressed by activated CD4+ T cells. IL-21 has a private receptor known as IL-21R, which is expressed on T, B, and NK cells. Within the B cell lineage, IL-21 regulates IgG1 production and cooperates with IL-4 for the production of multiple Ab classes in vivo. Initial studies have demonstrated that IL-21 has pleiotropic effects on the proliferation, differentiation, and effector functions of B, T, natural killer, and dendritic cells. Recombinant mouse IL-21 is a non-glycosylated protein, containing 130 amino acids, with a total molecular weight of 15.1 kDa.
Interleukin-21 Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Interleukin-21 in immunological assays.