Would you like to log in again?
rMouse Follistatin Protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
010001U88-0100 010001U88-0020 010001U88-1000 010001U88-0005
10801-454EA 885.78 USD InStock
10801-454 10802-938 10801-450 10802-942
rMouse Follistatin Protein, Rockland Immunochemicals
Proteins and Peptides
Recombinant Mouse Follistatin control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Follistatin is an autocrine acting protein that is expressed by many tissues, but at noteably higher levels in the ovary and skin. Follistatin functions to negatively regulate the signaling of a wide variety of TGF-beta family members (activin, BMPs, myostatin, GDF-11 and TGF-beta 1). Mechanistically, follistatin works as an antagonist by complexing with TGF-beta family members to prevent them from interacting with their signaling receptors. Recombinant mouse follistatin is a non-glycosylated protein, containing 289 amino acids, with a molecular weight of 31.6 kDa.
Follistatin Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Follistatin in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty