Would you like to log in again?
rMouse Activin A Protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
010001U62-1000 010001U62-0002 010001U62-0010 010001U62-0100
10801-828 10801-654 10801-834 10801-840
rMouse Activin A Protein, Rockland Immunochemicals
Recombinant Mouse Activin A Protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Activin A is a member of the TGFbeta family of proteins produced by many cell types throughout development. Activins interact with Type I and Type II serine/threonine kinases to signal to SMAD proteins to regulate a variety of functions, including cell proliferation, differentiation, wound healing, apoptosis, metabolism, etc. Activin A is a homodimer of two beta A chains and is not biologically active until the N terminal propeptide is cleaved from each. Mouse Activin A has 100% amino acid sequence identity to human, rat, porcine, bovine and feline proteins. Recombinant mouse Activin A is a non-glycosylated homodimer, containing two 117 amino acid chains, with a total molecular weight of 26.2 kDa.
Activin A Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Activin A in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty