Mouse EBI-3
Supplier: Rockland Immunochemicals
010-001-C55-1000 010-001-C55-0002 010-001-C55-0100
10801-222EA 960.74 USD
10801-222 10801-220 10801-224
Mouse EBI-3
Proteins and Peptides
Recombinant Mouse EBI-3 Protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Epstein-Barr Virus Induced Gene-3 (EBI3), is a secreted glycoprotein belonging to the hematopoietin receptor family and is related to the p40 subunit of IL-12. EBI3 was identified by its induced expression in B-lymphocytes in response to Epstein-Barr virus infection. EBI3 forms heterodimers with p28 to form IL-27 and with p35 to form IL-35. Both IL-27 and IL-35 have anti-inflammatory and regulatory activity. Recombinant mouse EBI3 is a non-glycosylated protein, containing 207 amino acids, with a molecular weight of 22.9 kDa.
Epstein-Barr Virus Induced Gene 3 Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Epstein-Barr Virus Induced Gene 3 in immunological assays. Product provided as lyophilized powder in 0.01M sodium citrate, pH 3.0,