Would you like to log in again?
rMouse EGF Protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
010001C26-0100 010001C26-1000
10802-104EA 143.05 USD InStock
10802-104 10801-246
rMouse EGF Protein, Rockland Immunochemicals
Proteins and Peptides
Recombinant Mouse EGF Protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Epidermal Growth Factor (EGF) is a growth factor that stimulates the proliferation of epithelial and epidermal cells. EGF family members are characterized by three intramolecular disulfide bonds and can bind to four different receptor tyrosine kinases known as EGFR/ErbB1, ErbB2, ErbB3, and ErbB4. Recombinant mouse EGF is a non-glycosylated protein, containing 54 amino acids, including 3 intra-molecular disulfide-bonds, with a molecular weight of 6.2 kDa.
Epidermal Growth Factor Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Epidermal Growth Factor in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty