Would you like to log in again?
rMouse IL-9 Protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
010001B96-1000 010001B96-0020 010001B96-0005 010001B96-0100
75934-766EA 7822.28 USD InStock
75934-766 75934-768 75929-180 75934-770
rMouse IL-9 Protein, Rockland Immunochemicals
Proteins and Peptides
Recombinant Mouse IL-9 control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Interleukin 9 (IL-9) is a cytokine produced by a subgroup of helper T cells, known as Th2 cells. IL-9 signals through its receptor IL-9R to induce cell proliferation and survival. Genetically, IL-9 has been identified as a gene associated with asthma. Recombinant mouse IL-9 is a non-glycosylated protein, containing 127 amino acids, with a molecular weight of 13.5 kDa.
Interleukin-9 Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Interleukin-9 in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty