Mouse IL-7
Supplier: Rockland Immunochemicals
010-001-B94-0005 010-001-B94-1000
75934-790EA 7868.42 USD
75934-790 75934-764
Mouse IL-7
Proteins and Peptides
Recombinant Mouse IL-7 control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Interleukin 7 (IL-7) is a hematopoietic cytokine that is constitutively produced by stromal cells from the bone marrow and thymus. IL-7 signals through the IL-7R to promote the differentiation of hematopoietic stem cells to lymphoid precursor cells, which give rise to T cells, B cell s, and natural killer cells. Human and mouse IL-7 show species cross-reactivity. Recombinant mouse IL-7 is a non-glycosylated protein, containing 172 amino acids, with a molecular weight of 17.4 kDa.
Interleukin-7 Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Interleukin-7 in immunological assays.