Mouse IL-4
Supplier: Rockland Immunochemicals
010-001-B93-0100 010-001-B93-0005 010-001-B93-0020 010-001-B93-1000
75929-172EA 139.92 USD
75929-172 75929-166 75929-168 75929-170
Mouse IL-4
Proteins and Peptides
Recombinant Mouse IL-4 control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Interleukin-4 (IL-4) is an immuno-modulatory cytokine, produced primarily by a subgroup of helper T cells, known as Th2. IL-4 is known to induce naïve helper T cells to differentiate to Th2 cells, which subsequently produce more IL-4. Th2 cells regulate a wide variety of other immune cells including, B cells, where it has the ability to promote IgG to IgE isotype switching. Recombinant mouse IL-4 is a non-glycosylated protein, containing 121 amino acids, with a molecular weight of 13.5 kDa.
Interleukin-4 Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Interleukin-4 in immunological assays.