Would you like to log in again?
MIP-1ß Mouse Recombinant Protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
010-001-B85
RL010-001-B85EA 311.76 USD InStock
RL010-001-B85
MIP-1ß Mouse Recombinant Protein, Rockland Immunochemicals
Proteins and Peptides
MIP-1ß Mouse Recombinant control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
MIP-1ß is a secreted monokine with inflammatory and chemokinetic properties and can exist as a homodimer. It belongs to the intercrine beta (chemokine CC) family. Recombinant Mouse Macrophage Inflammatory Protein-1 beta produced in E. coli is a single, non-glycosylated, polypeptide chain containing 69 amino acids and having a molecular mass of 7,808 Daltons.
MIP-1ß is suitable as a control for polyclonal or monoclonal anti-MIP-1ß in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty