Would you like to log in again?
IL-27/p28 Mouse Recombinant Protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
010-001B54-0.1
RL010001B540.1
IL-27/p28 Mouse Recombinant Protein, Rockland Immunochemicals
IL-27/p28 Mouse Recombinant control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Mouse IL-27/p28 Subunit, also known as Interleukin-30, is a member of the IL-12 family of cytokines. When combined with EBI3 (Epstein-Barr virus induced gene 3), the heterodimer formed is IL-27. Mouse p28 is a proinflammatory cytokine inducing immunomodulatory effects. Current research is underway to delineate specific biological functions. Recombinant mouse p28 is a single-chain 207 amino acid peptide with a mass of approximately 23,697 Daltons.
IL-27/p28 is suitable as a control for polyclonal or monoclonal anti-IL-27/p28 in immunological assays. Buffer Formulation: 10 mM sodium bicarbonate, pH 8.5.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty