Would you like to log in again?
MIP-3α Mouse Recombinant Protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
010-001B51-0.1
RL010001B510.1EA 697.16 USD InStock
RL010001B510.1
MIP-3α Mouse Recombinant Protein, Rockland Immunochemicals
Proteins and Peptides
MIP-3α Mouse Recombinant control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Recombinant Mouse MIP-3-alpha represents a CC chemokine that is expressed in the liver, lymph nodes, lung and other tissues and signals through the CCR6 receptor. Mouse MIP-3 alpha shows chemotactic activity towards lymphocytes and dendritic cells. Recombinant Mouse MIP-3 alpha has a molecular weight of 7.9 kDa, comprised of 70 amino acids.
MIP-3α is suitable as a control for polyclonal or monoclonal anti-MIP-3α in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty