Would you like to log in again?
IL-17F Mouse Recombinant Protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
010-001-B31
RL010-001-B31
IL-17F Mouse Recombinant Protein, Rockland Immunochemicals
IL-17F Mouse Recombinant control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Recombinant Mouse IL-17F produced in E.coli is a homodimeric, non-glycosylated polypeptide chain, the monomer containing a total of 133 amino acids and having a molecular mass of 14.9 kDa. Homodimeric interleukin-17F is a member of the IL-17 family of six homodimeric proteins (IL-17A-17F). Activated CD4+ T cells have also been shown to produce Mouse IL-17F. Activity studies reveal IL-17A to be the most potent, followed by IL-17A/F and IL-17F 100 fold lower than IL-17A. Recombinant Mouse IL-17F produced in E.Coli is a non-glycosylated polypeptide comprised of two monomeric subunit each of mIL-17F. The dimer consists of 266 amino acids, with an approximate molecular weight of 29.8 kDa.
IL-17F is suitable as a control for polyclonal or monoclonal anti-IL-17F in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty