Mouse IgA Kappa Isotype Control
Supplier: Rockland Immunochemicals
010-001-341
RL010-001-341EA 301.05 USD
RL010-001-341
Mouse IgA Kappa Isotype Control
Antibodies
Mouse IgA Kappa (κ) isotype Control
Mouse isotype controls are used in flow cytometry, western blot and ELISA and differentiate between immunoglobulin classes and subclasses. Isotype controls allow for the genetic variations or differences in the constant regions of the heavy and light chains. In mouse there are six relevant heavy chain isotypes and two light chain isotypes: heavy chain alpha - IgA, gamma - IgG 1, 2a, 2b, 3 and μ - IgM, light chain kappa and lambda.
Mouse IgA kappa isotype control can be utilized as a control or standard reagent in Flow cytometry, Western Blotting, and ELISA experiments where determination of sample isotype is important.
Applications: Control Protein, PCA Assays, ImmunoBlot