Mouse Recombinant Il1b
Supplier: Rockland Immunochemicals
010-001-319-0.1
RL0100013190.1EA 960.74 USD
RL0100013190.1
Mouse Recombinant Il1b
Proteins and Peptides
IL-1ß Mouse Recombinant control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Interleukin-1beta (IL-1β) is a proinflammatory cytokine produced in response to inflammatory agents by a variety of cells, including monocytes, macrophages, and dendritic cells (DCs). IL-1β and IL-1α are two distinct and independently regulated gene products that comprise IL-1 and signal through the Type 1 IL-1 receptor (IL-1R1). Although IL-1α is cell associated and IL-1β is secreted, they have nearly identical biological activity in that they induce adhesion molecule expression on epithelial cells, control fever induction, and play a role in arthritis and septic shock. Signaling activated by the IL-1R1 promotes these activities through a MYD88 signaling pathway similar to those associated with Toll receptors. Recombinant mouse IL-1β is a non-glycosylated single chain 17,394 Da protein containing 152 amino acid residues.
Interleukin-1beta (IL-1β) is suitable as a control for polyclonal or monoclonal anti-Interleukin-1beta (IL-1β) in immunological assays.