Would you like to log in again?
GM-CSF Mouse Recombinant Protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
010-001314-0.1
RL0100013140.1EA 739.47 USD InStock
RL0100013140.1
GM-CSF Mouse Recombinant Protein, Rockland Immunochemicals
Proteins and Peptides
GM-CSF Mouse Recombinant control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
M-GM-CSF is produced in response to a number of inflammatory mediators by mesenchymal cells present in the hemopoietic environment and at peripheral sites of inflammation. GM-CSF is able to stimulate the production of neutrophilic granulocytes, macrophages, and mixed granulocyte-macrophage colonies from bone marrow cells and can stimulate the formation of eosinophil colonies from fetal liver progenitor cells. GM-CSF can also stimulate some functional activities in mature granulocytes and macrophages. GM-CSF receptors shows significant homologies with other receptors for hematopoietic growth factors, including IL2 -beta, IL-3, IL-6, IL-7, EPO and the Prolactin receptors. Recombinant mouse GM- CSF produced in E.Coli is a single, non-glycosylated, polypeptide chain containing 125 amino acids and having a molecular mass of 14,285.35 Daltons.
GM-CSF is suitable as a control for polyclonal or monoclonal anti-GM-CSF in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty