Would you like to log in again?
Human Thrombopoietin Protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009F01W32-1000 009F01W32-0010 009F01W32-0002 009F01W32-0100
75932-866 75932-868 75932-872 75932-870
Human Thrombopoietin Protein, Rockland Immunochemicals
Recombinant Human Thrombopoietin control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Thrombopoietin (TPO) is a growth factor that is produced by liver and kidney tissue. TPO acts through the TPO receptor to promote megakaryocyte maturation and differentiation, which leads to the production of platelets. Recombinant human TPO is a non-glycosylated protein, containing the 174 amino acids of the receptor binding domain, with a molecular weight of 18.6 kDa.
Thrombopoietin Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Thrombopoietin in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty