Would you like to log in again?
Human TNF-alpha Protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009F01W31-0010 009F01W31-1000 009F01W31-0050
75933-212 75933-210 75933-214
Human TNF-alpha Protein, Rockland Immunochemicals
Recombinant Human TNF-alpha control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Tumor Necrosis Factor alpha (TNFα) is an inflammatory cytokine secreted by macrophages, monocytes, neutrophils, T cells, NK-cells following their stimulation by bacterial LPS. TNFα activates signals through two receptors, TNFR1, which is expressed on most cell types, and TNFR2, which is expressed mainly on immune cells. TNFα can have many functions including, to stimulate of phagocytosis in macrophages, to chemoattract neutrophils, to increase insulin resistance and to induce fever. Recombinant human TNF-α is a non-glycosylated protein, containing 158 amino acids, with a molecular weight of 17.5 kDa.
Tumor Necrosis Factor alpha Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Tumor Necrosis Factor alpha in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty