Would you like to log in again?
Human TGF-Beta3 Protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009F01W27-0100 009F01W27-0010 009F01W27-1000 009F01W27-0002
75933-082 75933-080 75933-078 75933-084
Human TGF-Beta3 Protein, Rockland Immunochemicals
Recombinant Human TGF-Beta3 control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
The Transforming Growth Factors (TGFs) are multifunctional peptides that regulate growth and differentiation in a variety of cells. Recent data suggests that individual TGF-β isoforms (TGF-β1, -β2 and –β3) have overlapping, yet distinct biological actions and target cell specificities, both in developing and adult tissues. TGF-β3 is a new isoform that is presumed to play an important role in wound repair and scarring. TGF-β3 is also thought to be involved in osteoblast proliferation, chemotaxis, and collagen synthesis. Recombinant human TGF-β3 is a non-glycosylated, disulfide-linked homodimer, containing two 112 amino acid chains, with a total molecular weight of 25.5 kDa.
Transforming Growth Factor beta 3 Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Transforming Growth Factor beta 3 in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty