Would you like to log in again?
Human PDGF-AB Protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009F01W10-0010 009F01W10-0002 009F01W10-0100 009F01W10-1000
75931-094 75931-098 75931-096 75931-092
Human PDGF-AB Protein, Rockland Immunochemicals
Recombinant Human PDGF-AB control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Platelet-Derived Growth Factor (PDGF) is a mitogenic peptide growth hormone carried in the alpha-granules of platelets and is released when platelets adhere to traumatized tissues. Connective tissue cells near the traumatized region respond by initiating the process of replication. The synthesis of PDGF can be induced by IL-1, IL-6, TNF-α, TGF-β and EGF. Recombinant human PDGF-AB is a non-glycosylated, disulfide-linked heterodimer, containing one 14.3 kDa alpha-chain and one 12.1 kDa beta-chain, with total molecular weight of 26.4 kDa.
Platelet Derived Growth Factor-AB Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Platelet Derived Growth Factor-AB in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty