Would you like to log in again?
Human IL-21 Protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009F01V61-0002 009F01V61-1000 009F01V61-0100 009F01V61-0010
75929-130EA 139.57 USD InStock
75929-130 75929-124 75929-128 75929-126
Human IL-21 Protein, Rockland Immunochemicals
Proteins and Peptides
Recombinant Human IL-21 control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Interleukin-21 (IL-21) is a member of the common gamma-chain family of cytokines (includes IL-2, IL-4, IL-7, IL-9, IL-13, and IL-15) and is expressed by activated CD4+ T cells. IL-21 has a private receptor known as IL-21R, which is expressed on T, B, and NK cells. Within the B cell lineage, IL-21 regulates IgG1 production and cooperates with IL-4 for the production of multiple Ab classes in vivo. Initial studies have demonstrated that IL-21 has pleiotropic effects on the proliferation, differentiation, and effector functions of B, T, natural killer, and dendritic cells. Recombinant human IL-21 is a non-glycosylated protein, containing 133 amino acids, with a total molecular weight of 15.5 kDa.
Interleukin-21 Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Interleukin-21 in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty