Would you like to log in again?
Human GM-CSF Protein , Rockland Immunochemicals
Supplier: Rockland Immunochemical
009F01V11-0100 009F01V11-0005 009F01V11-0020 009F01V11-1000
75928-198EA 565.01 USD InStock
75928-198 75928-202 75928-200 75928-196
Human GM-CSF Protein , Rockland Immunochemicals
Proteins and Peptides
Recombinant Human GM-CSF control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Granulocyte Macrophage Colony Stimulating Factor (GM-CSF) is hematopoietic factor produced by endothelial cells, monocytes, fibroblasts and T cells in response to a number of inflammatory mediators. GM-CSF is able to stimulate the production of neutrophilic granulocytes, macrophages, and mixed granulocyte-macrophage colonies from bone marrow cells. GM-CSF can also stimulate some functional activities in mature granulocytes and macrophages. Human and mouse GM-CSF show no cross-reactivity. Recombinant human GM-CSF is a non-glycosylated protein, containing 128 amino acids, with a molecular weight of 14.6 kDa.
Granulocyte Macrophage Colony Stimulating Factor Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Granulocyte Macrophage Colony Stimulating Factor in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty