Would you like to log in again?
Human IL-3 Protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009F01B92-0002 009F01B92-1000
75929-146 75928-810
Human IL-3 Protein, Rockland Immunochemicals
Recombinant Human IL-3 control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Interleukin-3 (IL-3) is a cytokine produced by activated T cells and mast cells. IL-3 is able to induce the differentiation of hematopoietic stem cells to precursor cells of myeloid lineage (erythrocytes, megakaryocytes, granulocytes, dendritic cells and monocytes). IL-3 also has functions in the nervous system and appears to be important in several chronic inflammatory diseases. Recombinant human IL-3 is a non-glycosylated protein, containing 133 amino acids, with a molecular weight of 15 kDa.
Human Interleukin-3 Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Human Interleukin-3 in immunological assays. Buffer formation: 10 mM sodium phosphate, 50 mM sodium sulfate, pH 7.5.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty