Would you like to log in again?
HUMAN IgA, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-0106-0001
75928-824EA 184.58 USD InStock
75928-824
HUMAN IgA, Rockland Immunochemicals
Proteins and Peptides
Human IgA whole molecule
  • control or standard reagent
  • SDS, Western Blotting, and ELISA experiments
Immunoglobulin A is the primary responder in muscosal immunity, and comprises close to 75% of the total immunoglobulin produced. IgA can also be secreted and is protected from degradation by many proteolytic enzymes (which allows it to be secreted along the gastrointestinal tract). Immunoglobulin A only weakly activates the complement system and is not readily opsonized.
Human IgA whole molecule can be utilized as a control or standard reagent in SDS, Western Blotting and ELISA experiments.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty