Would you like to log in again?
Human IgG whole molecule, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-0102
RL009-0102-10EA 158.86 USD InStock
RL009-0102-10
Human IgG whole molecule, Rockland Immunochemicals
Proteins and Peptides
Human IgG whole molecule
  • control or standard reagent
  • SDS, Western Blotting, and ELISA experiments
Human IgG purified protein (Immunoglobulin G) are antibody molecules. Human IgG is composed of four peptide chains — two heavy chains gamma and two light chains. Human IgG has two antigen binding sites. Other Immunoglobulins may be described in terms of polymers with the IgG structure considered the monomer. Human IgG typically constitutes 75% of serum immunoglobulins. Human IgG molecules are synthesized and secreted by plasma B cells.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty