Would you like to log in again?
HUMAN IgM PE, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-008-007
RL009-008-007EA 179.21 USD InStock
RL009-008-007
HUMAN IgM PE, Rockland Immunochemicals
Proteins and Peptides
Human IgM whole molecule Phycoerythrin conjugated
  • control reagent
  • Flow Cytometry, Immunohistochemistry, and Western Blotting
Immunoglobulin M is the largest antibody isotype and the first to be secrected against an initial exposure to antigen. IgM is predominantly produced in the spleen. Formed from covalently linking 5 immunoglobulins together, the approixmate molecular weight of IgM is 900kDa and possesses 10 binding sites (though due to the size of most antigens, not all sites are capable of binding at once). Due to this large size, IgM is typically isolated to the serum.
Human IgM whole molecule Phycoerythrin conjugated can be utilized as a control reagent in Flow Cytometry, Immunohistochemistry, and Western Blotting. Researchers should determine optimal titers for applications that are not stated.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty