Human TRAIL Protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-W34-1000 009-001-W34-0050 009-001-W34-0010
75933-038EA 2178.59 USD
75933-038 75933-040 75933-042
Human TRAIL Protein, Rockland Immunochemicals
Proteins and Peptides
Recombinant Human TRAIL control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
TNF-Related Apoptosis-Inducing Ligand (TRAIL) is produced by a wide variety of cell types and is shown to be a cytotoxic protein that induces apoptosis in tumor cells through activation of the death receptors, DR4 and DR5. Human TRAIL is active on mouse cells. Recombinant human TRAIL is a non-glycosylated protein, containing the 168 amino acid extracellular portion of TRAIL that is homologous to TNF. It has a molecular weight of 19.5 kDa.
TRAIL Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-TRAIL in immunological assays. Product is provided as lyophilized powder in 0.05M Sodium Chloride and 0.01M Sodium Phosphate, pH 7.5.