Would you like to log in again?
Human TGF-Beta1 Protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009001W26-0001
75933-056
Human TGF-Beta1 Protein, Rockland Immunochemicals
Recombinant Human TGF-Beta1 control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Transforming Growth Factor-beta 1 (TGF-β1) is a member of the TGF-beta superfamily. Members of this family are known to exhibit regulatory activity in immunity and proliferation pathways. TGF-β1 signals through SMAD proteins via the TGF-β1 RI and RII receptors. Mature human TGF-β1 is a glycosylated homodimer, containing two 112 amino acids chains, with a total molecular weight of 25 kDa.
Transforming Growth Factor beta 1 Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Transforming Growth Factor beta 1 in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty