Would you like to log in again?
Human Sonic Hedgehog Protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009001W24-0005
75932-114EA 135.05 USD InStock
75932-114
Human Sonic Hedgehog Protein, Rockland Immunochemicals
Proteins and Peptides
Recombinant Human Sonic Hedgehog control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Sonic hedgehog (SHH) is a member of a small group of secreted proteins that are essential for development in both vertebrates and invertebrates. Three mammalian hedgehog genes (sonic, desert, Indian) share about 60% homology and all signal via the same receptors. Recombinant human SHH is a non-glycosylated protein, containing 175 amino acids, with a molecular weight of 19.7 kDa. The Cys at position 25 has been substituted with Ile.
Sonic Hedgehog Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Sonic Hedgehog in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty