Would you like to log in again?
Human RELM beta Protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009001W18-0100 009001W18-0005 009001W18-0025 009001W18-1000
75931-520EA 565.01 USD InStock
75931-520 75931-528 75931-524 75931-516
Human RELM beta Protein, Rockland Immunochemicals
Proteins and Peptides
Recombinant Human RELM beta control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Resistin-Like Molecule-beta (RELM- β) is a member of a recently identified family of secreted proteins containing conserved cysteines in their C terminus. The RELM family consists of Resistin (also called FIZZ3), RELM-α (FIZZ1), and RELM-γ. Only Resistin and RELM-β were found in humans whereas all four RELM family members have been identified in rodents. Recombinant human RELM-β is a non-glycosylated, disulfide-linked homodimer, containing two 89 amino acid chains, with a total molecular weight of 19 kDa.
RELM beta Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-RELM beta in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty