Would you like to log in again?
Human LIF Protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009001V72-0025 009001V72-0100 009001V72-0005 009001V72-1000
75933-808EA 307.26 USD InStock
75933-808 75934-850 75933-810 75934-846
Human LIF Protein, Rockland Immunochemicals
Proteins and Peptides
Recombinant Human LIF control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Human Leukemia Inhibitory Factor (LIF) is member of the IL-6 family and is made by a variety of tissues. Human LIF signals through gp130 to activiate STAT3 in human ESCs . In mouse ESCs, LIF promotes self-renewal and pluripotentcy, but in human cells there is no indication that LIF has the same function. Instead, FGF-basic (FGF-2) and Activin A are used to promote self-renewal and pluripotent potential. Recombinant human LIF is a non-glycosylated protein, containing 181 amino acids, with a molecular weight of 19.9 kDa.
Leukemia Inhibitory Factor Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Leukemia Inhibitory Factor in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty