Would you like to log in again?
Human IL-19 Protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009001V58-0002 009001V58-1000 009001V58-0010 009001V58-0100
75934-716EA 135.05 USD InStock
75934-716 75929-102 75929-104 75929-106
Human IL-19 Protein, Rockland Immunochemicals
Proteins and Peptides
Recombinant Human IL-19 control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Interleukin-19 (IL-19) is a member of the IL-10 family produced by resting B cells and monocytes. IL-19 is thought to be up-regulated when monocytes are treated with GM-CSF or LPS. IL-19 shares a receptor complex with IL-20, and acts to induce IL-6 and TNF-α production by monocytes and promotes the Th2 response. Recombinant human IL-19 is a non-glycosylated protein, containing 154 amino acids, with a molecular weight of 17.9 kDa.
Interleukin-19 Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Interleukin-19 in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty