Would you like to log in again?
Human IL-13 Protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009001V52-0002 009001V52-1000 009001V52-0010 009001V52-0100
75934-674EA 126.75 USD InStock
75934-674 75929-054 75929-058 75934-670
Human IL-13 Protein, Rockland Immunochemicals
Proteins and Peptides
Recombinant Human IL-13 control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Interleukin-13 (IL-13) is an important cytokine secreted from Th2 cells. The functions attributed to IL-13 overlap significantly with those of IL-4 (induces IgE secretion from B cells and inhibits expression of inflammatory cytokines such as IL-1β, TNF-α, IL-8 and IL-6), but differs from IL-4 in that IL-13 seems to link inflammatory responses of immune cells to the pathophysiological changes in the surrounding non-immune cells. The receptor subunits of IL-13 consist of IL-4Rα, IL-13Rα1 and IL-13Rα2. Human and mouse IL-13 are cross-reactive. Recombinant human IL-13 is a non-glycosylated variant of native IL-13, containing 115 amino acids (additional Gln at position 113), with a molecular weight of 12.6 kDa.
Interleukin-13 Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Interleukin-13 in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty