Would you like to log in again?
Human Growth Hormone Protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009001V13-0100 009001V13-0500 009001V13-5000
75928-266EA 139.57 USD InStock
75928-266 75928-270 75928-268
Human Growth Hormone Protein, Rockland Immunochemicals
Proteins and Peptides
Recombinant Human Growth Hormone control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Growth Hormone (GH) is an important growth factor made in and secreted throughout the body by the anterior pituitary gland. It stimulates growth and cell reproduction and regeneration in humans and other animals. The inability to appropriately produce or respond to GH results in diseases of decreased stature in children and adults. Although GH is used to treat several growth disorders, it is thought to be a very complex hormone with many of its functions yet to be uncovered. Recombinant human GH is non-glycosylated protein, containing 192 amino acids, with a molecular weight of 22.2 kDa.
Growth Hormone Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Growth Hormone in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty