Would you like to log in again?
Human GDF-5 Protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009001U94-0050 009001U94-0010
75928-264EA 307.26 USD InStock
75928-264 75928-262
Human GDF-5 Protein, Rockland Immunochemicals
Proteins and Peptides
Recombinant Human GDF-5 control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Growth/Differentiation Factor 5 (GDF-5) is a growth factor that regulates cell proliferation and differentiation in embryonic and adult tissues. GDF-5 is part of the TGF family of proteins and is closely related to the BMP family of proteins. Recombinant human GDF-5 is a non-glycosylated homodimer, containing two 120 amino acids chains, with a total molecular weight of 27.4 kDa. To enable bacterial expression the N-terminal sequence of Ala-Pro-Leu-Thr was replaced with a Lys.
Growth and Differentiation Factor-5 Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Growth and Differentiation Factor-5 in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty