Human Recombinant FGF147 basic
Supplier: Rockland Immunochemicals
009-001-U82-0010 009-001-U82-0050 009-001-U82-1000
10802-818EA 2178.59 USD
10802-818 10802-820 10802-822
Human Recombinant FGF147 basic
Proteins and Peptides
Recombinant Human FGF147 basic control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Fibroblast Growth Factors (FGFs) are a 22 member family of proteins known to be involved in angiogenesis, wound healing and embryonic development. As a family, they bind to heparin and signal through four receptor tyrosine kinases called, FGFR1, 2, 3 and 4. Although the mechanism remains unclear, FGF-basic 147 (variant of FGF basic 154), also called FGF-2, is a critical component in keeping embryonic stem cells undifferentiated in cell culture systems. Recombinant human FGF-b 147 (FGF-2) is a non-glycosylated protein, containing 147 amino acids, with a molecular weight of 16.5 kDa.
Fibroblast Growth Factor 147 basic Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Fibroblast Growth Factor 147 basic in immunological assays.