Human Recombinant FGF-21
Supplier: Rockland Immunochemicals
009-001-U77-0020 009-001-U77-1000 009-001-U77-0100 009-001-U77-0005
10801-374EA 4838.75 USD
10801-374 10802-858 10802-856 10802-860
Human Recombinant FGF-21
Proteins and Peptides
Recombinant Human FGF-21 control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Fibroblast Growth Factors (FGFs) are a 22 member family of proteins known to be involved in angiogenesis, wound healing and embryonic development. As a family, they bind to heparin and signal through four receptor tyrosine kinases called, FGFR1, 2, 3 and 4. FGF-21 is most closely related to FGF-19 and shows low binding to heparin and is highly diffusible. Recombinant human FGF-21 is a non-glycosylated protein, containing 182 amino acids, with a molecular weight of 19.5 kDa.
Fibroblast Growth Factor-21 Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Fibroblast Growth Factor-21 in immunological assays. Lyophilized in 0.1M Sodium Chloride and 0.01M Sodium Phosphate, pH 7.5.