Would you like to log in again?
Human BAFF - Protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009001U65-0100 009001U65-1000 009001U65-0005 009001U65-0020
10801-970 10801-968 10801-974 10801-972
Human BAFF - Protein, Rockland Immunochemicals
Recombinant Human BAFF - control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Human B cell Activating Factor (BAFF), is a Type II member of the TNF superfamily. Although it is typically a transmembrane protein expressed on T cells, macrophages and dendritic cells, it can also be cleaved in the extracellular region to produce a soluble form detectable in the serum. BAFF is thought to be important for the survival and maturation of peripheral B cells. BAFF initiates signaling through three receptors with human BAFF having activity on mouse splenocytes. Recombinant human BAFF is a non-glycosylated protein containing a N terminal His tag, a thrombin cut site and 153 amino acids of the TNF-like domain. This protein has a total molecular weight of 18.5 kDa.
Human BAFF Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-BAFF in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty