Human Activin A Protein, Rockland Immunochemicals
009001U62-0010 009001U62-1000 009001U62-0002 009001U62-0100
10801-838EA 732.02 USD
10801-838 10801-826 10801-832 10801-652
Human Activin A Protein, Rockland Immunochemicals
Recombinant Human Activin A control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Activin A is a member of the TGF beta family of proteins produced by many cell types throughout development. Activins interact with Type I and Type II serine/threonine kinases to signal to SMAD proteins to regulate a variety of functions, including cell proliferation, differentiation, wound healing, apoptosis, metabolism, etc. Activin A is a homodimer of two beta A chains and is not biologically active until the N terminal propeptide is cleaved from each. Human Activin A has 100% amino acid sequence identity to mouse, rat, porcine, bovine and feline proteins. Recombinant human Activin A is a non-glycosylated homodimer, containing two 117 amino acid chains, with a total molecular weight of 26.2 kDa.
Activin A Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Activin A in immunological assays.