Would you like to log in again?
XIAP protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-U61S 009-001-U61
75935-498EA 353.41 USD InStock
75935-498 75933-652
XIAP protein-GST fusion, Rockland Immunochemicals
Proteins and Peptides
XIAP control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
XIAP is a member of a family of proteins which inhibit apoptosis and act as mammalian cell-death suppressors. XIAP inhibits at least 2 members of the caspase family of cell-death proteases, caspase-3 and caspase-7 (1). In addition, XIAP inhibits apoptosis induced by menadione, a potent inducer of free radicals, and ICE. Furthermore, XIAP can mediate protection of cells from apoptosis by utilizing both a JNK1 activation pathway that involves ILPIP and a caspase inhibition pathway that is independent of ILPIP (2). Disruption of the XIAP gene in human colon cancer cells can enhance sensitivity of these cells to exogenously added TRAIL suggesting that XIAP is a nonredundant modulator of TRAIL-mediated apoptosis. XIAP Protein is ideal for investigators involved in Signaling Proteins, Apoptosis Proteins, Apoptosis/Autophagy, Cancer, Inflammation, and Neurobiology research.
XIAP Protein is stored in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. XIAP Protein is suitable for use in Western Blot. Expect a band approximately ~84kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty