Would you like to log in again?
WWP2 protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-U60S 009-001-U60
75933-646EA 353.41 USD InStock
75933-646 75933-648
WWP2 protein-GST fusion, Rockland Immunochemicals
Proteins and Peptides
WWP2 control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
WWP2 or WW domain containing E3 ubiquitin protein ligase 2 is a member of the Nedd4 family of E3 ligases, which play an important role in protein ubiquitination. WWP2 contains four WW domains and may play a role in multiple processes including chondrogenesis and the regulation of oncogenic signaling pathways via interactions with Smad proteins and the tumor suppressor PTEN. WWP2 is an E3 ubiquitin ligase that targets transcription factor Oct-4 for ubiquitination (1). WWP2 which is also called AIP2 can bind to atrophin-1 (2). WWP2 Protein is ideal for investigators involved in Signaling Proteins, Ubiquitin Proteins, Cancer, Cardiovascular Disease, and Cell Cycle research.
WWP2 Protein is stored in 50mM Tris-HCl, pH 7.5, 50mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. WWP2 Protein is suitable for use in Western Blot. Expect a band approximately ~130kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty