Would you like to log in again?
VRK3 protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-U58S 009-001-U58
75933-598EA 353.41 USD InStock
75933-598 75933-600
VRK3 protein-GST fusion, Rockland Immunochemicals
Proteins and Peptides
VRK3 control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
VRK3 is a member of the vaccinia-related kinase (VRK) family of serine/threonine protein kinases which has substitutions at several residues within the ATP binding motifs that in other kinases have been shown to be required for catalysis. Profound changes to the active site region underlie the loss of catalytic activity of VRK3 since it cannot bind ATP because of residue substitutions in the binding pocket (1). VRK3 is widely expressed in human tissues and the protein localizes to the nucleus. VRK3 regulates several transcription factors, nuclear envelope assembly, and chromatin condensation and is also required for cell cycle progression (2). VRK3 Protein is ideal for investigators involved in Signaling Proteins, Cellular Proteins, Apoptosis/Autophagy, Cancer, ERK/MAPK Pathway, JNK/SAPK Pathway, p38 Pathway, and Ser/Thr Kinases research.
VRK3 Protein is stored in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. VRK3 Protein is suitable for use in Western Blot. Expect a band approximately ~80kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty