Would you like to log in again?
VLK protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-U57S 009-001-U57
75933-586EA 353.41 USD InStock
75933-586 75933-588
VLK protein-GST fusion, Rockland Immunochemicals
Proteins and Peptides
VLK control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
VLK or vertebrate lonesome kinase is a protein kinase which is first expressed in E-cadherin-positive anterior visceral endoderm and mesendoderm, and later its expression is confined to E-cadherin-negative mesenchyme. VLK regulates the rate of protein export from the Golgi and thereby plays an important role in the formation of functional stroma by mesenchymal cells (1). Targeted disruption of VLK leads to a defect in lung development and delayed ossification of endochondral bone. VLK deficient mice display neonatal lethality due to respiratory failure, with a suckling defect arising from a cleft palate. VLK is required for the appropriate timing of flat proliferative chondrocyte differentiation (2). VLK Protein is ideal for investigators involved in Signaling Proteins, Cellular Proteins, Cancer, Cytoplasmic Tyrosine Kinases, Invasion/Metastasis, Neurobiology, and Ser/Thr Kinases research.
VLK Protein is stored in 50mM 50mM Tris-HCl, pH 7.5, 50mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. VLK Protein is suitable for use in Western Blot. Expect a band approximately ~69kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty