Would you like to log in again?
UCHL5 protein-HIS Epitope, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-U52 009-001-U52S
75933-414EA 307.26 USD InStock
75933-414 75933-412
UCHL5 protein-HIS Epitope, Rockland Immunochemicals
Proteins and Peptides
UCHL5 control protein-HIS Epitope
  • Control Protein
  • PCA Assays
  • ImmunoBlot
UCHL5 or ubiquitin carboxyl-terminal hydrolase L5 is a member of the UCH enzyme family which contains a conserved catalytic domain and a nonconserved extended C-terminal tail. UCHL5 overexpression increases TGFbR1-dependent transcription. Knockdown of UCHL5 expression by RNA interference abrogated TGF-beta-dependent transcription (1). UCHL5 is a component of the 26S proteasome that functions in editing polyubiquitinated protein substrates (2). The activity of the C-terminal hydrolase UCHL5 is upregulated in a majority of tumor tissues compared to the adjacent normal tissues. UCHL5 Protein is ideal for investigators involved in Signaling Proteins, Angiogenesis, Apoptosis/Autophagy, Cancer, Cell Cycle, Cellular Stress, Inflammation, Invasion/Metastasis, and Neurobiology research.
UCHL5 Protein is stored in 50mM sodium phosphate, pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, 25% glycerol. UCHL5 Protein is suitable for use in Western Blot. Expect a band approximately ~38kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty